missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H8 Antibody (OTI4H5), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74908AF700
This item is not returnable.
View return policy
Description
ZC3H8 Monoclonal specifically detects ZC3H8 in Human samples. It is validated for Immunohistochemistry.
Specifications
| ZC3H8 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ZC3H8 | |
| Full length recombinant protein of human ZC3H8 (NP_115883) produced in E.coli. | |
| 0.1 mL | |
| 84524.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI4H5 | |
| Immunohistochemistry | |
| Fliz1, ZC3HDC8, zinc finger CCCH domain-containing protein 8, zinc finger CCCH type domain containing 8, zinc finger CCCH-type containing 8, zinc finger CCCH-type domain containing 8 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction