missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H7A Antibody (PCRP-ZC3H7A-1D6), Janelia Fluor™ 646, Novus Biologicals™
Shop All Bio Techne ProductsDescription
ZC3H7A Monoclonal antibody specifically detects ZC3H7A in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | ZC3H7A |
| Applications | Flow Cytometry, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | PCRP-ZC3H7A-1D6 |
| Conjugate | Janelia Fluor 646 |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | FLJ10027, FLJ20318, HSPC055, ZC3H7, ZC3HDC7, zinc finger CCCH domain-containing protein 7A, zinc finger CCCH type domain containing 7, zinc finger CCCH-type containing 7, zinc finger CCCH-type containing 7A, zinc finger CCCH-type domain containing 7, zinc-finger protein AY163807 |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human ZC3H7A protein (Uniprot: Q8IWR0) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?