missing translation for 'onlineSavingsMsg'
Learn More

ZC3H7A Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18346364 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346364 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18346364 Supplier Novus Biologicals Supplier No. NBP306301AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZC3H7A Polyclonal specifically detects ZC3H7A in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3H7A
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias FLJ10027, FLJ20318, HSPC055, ZC3H7, ZC3HDC7, zinc finger CCCH domain-containing protein 7A, zinc finger CCCH type domain containing 7, zinc finger CCCH-type containing 7, zinc finger CCCH-type containing 7A, zinc finger CCCH-type domain containing 7, zinc-finger protein AY163807
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ZC3H7A fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29066
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.