missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20041-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody recognizes near the C-term of ZC3H4.
This gene encodes a member of a family of CCCH (C-x8-C-x5-C-x3-H type) zinc finger domain-containing proteins. These zinc finger domains, which coordinate zinc finger binding and are characterized by three cysteine residues and one histidine residue, are nucleic acid-binding. Other family members are known to function in post-transcriptional regulation.Specifications
| ZC3H4 | |
| Polyclonal | |
| Unconjugated | |
| ZC3H4 | |
| C19orf7, KIAA1064, ZC3H4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23211, 330474 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6ZPZ3, Q9UPT8 | |
| Zc3h4 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction