missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-03341
This item is not returnable.
View return policy
Description
ZC3H3 Polyclonal specifically detects ZC3H3 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| ZC3H3 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| KIAA0150zinc finger CCCH-type domain containing 3, ZC3HDC3, zinc finger CCCH domain-containing protein 3, zinc finger CCCH type domain containing 3, zinc finger CCCH-type containing 3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 23144 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:1000, Immunoprecipitation 2-5 μg/mg lysate | |
| NP_055932.2 | |
| ZC3H3 | |
| The immunogen recognized by this antibody maps to a region between residue 898 and 948 of human zinc finger CCCH-type containing 3 using the numbering given in entry NP_055932.2 (GeneID 23144). | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction