missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZC3H15 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA556637

Product Code. 15794541

  • £485.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: IVCKHFLEAI ENNKYGWFWV CPGGGDICMY RHALPPGFVL KKDKKKEEKE DEISLEDLIE RERSALGPNV TKITL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

Protects DRG1 from proteolytic degradation (PubMed:19819225). Stimulates DRG1 GTPase activity likely by increasing the affinity for the potassium ions (PubMed:23711155). [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZC3H15
Polyclonal
Unconjugated
Zc3h15
1700006A17Rik; 1810012H02Rik; 2610312B22Rik; brain zinc finger protein; DFRP1; DRG family-regulatory protein 1; epo-immediate response gene protein FM22; FM22; HSPC303; HT010; Ierepo4; LEREPO4; likely ortholog of mouse immediate early response erythropoietin 4; MSTP012; p48ZnF; PP730; Zc3h15; Zinc finger CCCH domain-containing protein 15; zinc finger CCCH-type containing 15
Rabbit
Antigen affinity chromatography
RUO
55854
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.4 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8WU90
Zc3h15
Recombinant protein corresponding to Human ZC3H15. Recombinant protein control fragment (Product #RP-95966).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.