Learn More
ZC3H14 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28190-1-AP-20UL
This item is not returnable.
View return policy
Description
The ZC3H14 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ZC3H14. This antibody recognizes human, mouse antigen. The ZC3H14 antibody has been validated for the following applications: ELISA analysis.
Specifications
| ZC3H14 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 79882 | |
| IgG |
| ELISA | |
| 233 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |