Learn More
Invitrogen™ ZC3H14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587844
Description
Immunogen sequence: MRMSSKFPSP PLPIFLPPEP VDLGSITSSS CSLNELDNIS HLLRKISADI NEIKGMKAAI LTVEANLFDL NVRVSKNEAK ISSLEVKMNE YSTTYECNRQ FEDQEEDTES QSRTTDVKII GFLRNVEKGT QQRQLLSRLQ IDPVMAETLQ MSQAEMSELS VAQKPEKLLE RCKYWPACKN GDECAYHHPI SPCKAFPNCK FAEKCLFVHP NCKYDAKCTK PDCPFTHVSR RIPVLSPKPV APPAPPSSSQ LCRYFPACKK MECPFYHPKH CRFNTQCTRP DCTFYHPTIN VPPRHALKWI RPQTSE; Positive Samples: MCF7, A-549, HT-1080, 293T; Cellular Location: Cytoplasm, Nucleus speckle.
ZC3H14 belongs to the ZC3H14 family.It contains 2 C3H1-type zinc fingers. ZC3H14 binds the polyadenosine RNA oligonucleotides.
Specifications
| ZC3H14 | |
| Polyclonal | |
| Unconjugated | |
| ZC3H14 | |
| 1010001P15Rik; 1700016A15Rik; 2700069A02Rik; AU014748; FLJ11806; Mammalian suppressor of tau pathology-2; MSUT-2; Npuk68; nuclear protein UKp68; nuclear protein UKp83/UKp68; NY-REN-37; Renal carcinoma antigen NY-REN-37; SUT2; UKp68; Zc3h14; Zinc finger CCCH domain-containing protein 14; zinc finger CCCH type containing 14; zinc finger CCCH-type containing 14 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79882 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.01 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6PJT7 | |
| ZC3H14 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-306 of human ZC3H14 (NP_9975452). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.