missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H14 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93461-0.1ml
This item is not returnable.
View return policy
Description
ZC3H14 Polyclonal antibody specifically detects ZC3H14 in Human samples. It is validated for Western Blot, ELISA
Specifications
| ZC3H14 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA | |
| FLJ11806, Mammalian suppressor of tau pathology-2, MGC26892, MSUT-2, nuclear protein UKp68, NY-REN-37, Renal carcinoma antigen NY-REN-37, SUT2, UKp68, zinc finger CCCH domain-containing protein 14, zinc finger CCCH-type containing 14 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-306 of human ZC3H14 (NP_997545.2). MRMSSKFPSPPLPIFLPPEPVDLGSITSSSCSLNELDNISHLLRKISADINEIKGMKAAILTVEANLFDLNVRVSKNEAKISSLEVKMNEYSTTYECNRQFEDQEEDTESQSRTTDVKIIGFLRNVEKGTQQRQLLSRLQIDPVMAETLQMSQAEMSELSVAQKPEKLLERCKYWPACKNGDECAYHHPISPCKAFPNCKFAEKCLFVHPNCKYDAKCTKPDCPFTHVSRRIPVLSPKPVAPPAPPSSSQLCRYFPACKKMECPFYHPKHCRFNTQCTRPDCTFYHPTINVPPRHALKWIRPQTSE | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79882 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction