Learn More
ZC3H13 Mouse, anti-Human, Clone: 3E8C5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68526-1-IG-150UL
This item is not returnable.
View return policy
Description
The ZC3H13 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ZC3H13. This antibody recognizes human antigen. The ZC3H13 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ZC3H13 | |
| 3E8C5 | |
| DKFZp434D1812, FLJ35669, KIAA0853, ZC3H13 | |
| 150 μL | |
| 23091 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG2b |