missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H12D Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24991-1-AP-150UL
This item is not returnable.
View return policy
Description
ZC3H12D, also known as MCPIP4, is a member of a family of novel CCCH-zinc finger proteins that includes ZC3H12A, a protein that is thought to be involved in macrophage activation, host immunity and inflammatory diseases. ZC3H12D is highly expressed in spleen, intestine, and colon. Similar to ZC3H12A and ZC3H12C, ZC3H12D expression in murine macrophages is also increased after treatment with lipopolysaccharide (LPS), although not to the same level as ZC3H12A. This pattern of expression was not duplicated in human-derived macrophages, suggesting it also may play a different role in host immunity and inflammatory response.Specifications
| ZC3H12D | |
| Polyclonal | |
| Unconjugated | |
| ZC3H12D | |
| C6orf95, MCP induced protein 4, MCPIP4, p34, Probable ribonuclease ZC3H12D, TFL, ZC3H12D | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 340152 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| A2A288 | |
| ZC3H12D | |
| ZC3H12D Fusion Protein Ag21770 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction