missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H12D Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98171G
This item is not returnable.
View return policy
Description
ZC3H12D Polyclonal antibody specifically detects ZC3H12D in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ZC3H12D | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| C6orf95, FLJ46041, MCP-induced protein 4, p34, TFL, transformed follicular lymphoma, tumor suppressor TFL, zinc finger CCCH-type containing 12D | |
| Produced in rabbits immunized with E. coli-derived Human ZC3H12D fragment. | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 340152 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction