missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H12C Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Description
ZC3H12C Polyclonal antibody specifically detects ZC3H12C in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
Specifications
Specifications
| Antigen | ZC3H12C |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Formulation | PBS, 50% glycerol, pH7.3 |
| Gene Alias | KIAA1726, MCP induced protein 3, MCP-induced protein 3, MCPIP3, probable ribonuclease ZC3H12C, zinc finger CCCH domain-containing protein 12C, zinc finger CCCH-type containing 12C |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 735-883 of human ZC3H12C (NP_203748.1). KAPHLGRSLVATRIDSISDSRLYDSSPSRQRKPYSRQEGLGSWERPGYGIDAYGYRQTYSLPDNSTQPCYEQFTFQSLPEQQEPAWRIPYCGMPQDPPRYQDNREKIYINLCNIFPPDLVRIVMKRNPHMTDAQQLAAAILVEKSQLGY |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?