missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZC3H12A Polyclonal Antibody
GREENER_CHOICE

Product Code. 12859072 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12859072 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12859072 Supplier Invitrogen™ Supplier No. PA520926

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is Raji cell lysate. PA5-20926 can be used with blocking peptide PEP-1040.

ZC3H12A is an MCP1 -induced protein that acts as a transcriptional activator and causes cell death of cardiomyocytes, possibly via induction of genes associated with apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3H12A
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ZC3H12A
Gene Accession No. Q5D1E7, Q5D1E8
Gene Alias BC036563; bifunctional endoribonuclease and deubiquitinase ZC3H12A; dJ423B22.1; Endoribonuclease ZC3H12A; MCP-1 treatment-induced protein; MCP-induced protein 1; MCPIP; MCPIP1; Monocyte chemotactic protein-induced protein 1; Reg1; Regnase-1; RGD:1306776}; RGD1306776; ribonuclease ZC3H12A; RP3-423B22.1; zc3h12a; zc3h12a {ECO:0000312; Zinc finger CCCH domain-containing protein 12A; zinc finger CCCH type containing 12A; zinc finger CCCH-type containing 12A
Gene Symbols ZC3H12A
Host Species Rabbit
Immunogen A 17 amino acid peptide near the center of human ZC3H12A.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230738, 80149
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.