missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZC3H12A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524458
This item is not returnable.
View return policy
Description
ZC3H12A Polyclonal Antibody for Flow, IHC (P), Western Blot
ZC3H12A is an MCP1 -induced protein that acts as a transcriptional activator and causes cell death of cardiomyocytes, possibly via induction of genes associated with apoptosis.
Specifications
| ZC3H12A | |
| Polyclonal | |
| Unconjugated | |
| ZC3H12A | |
| BC036563; bifunctional endoribonuclease and deubiquitinase ZC3H12A; dJ423B22.1; Endoribonuclease ZC3H12A; MCP-1 treatment-induced protein; MCP-induced protein 1; MCPIP; MCPIP1; Monocyte chemotactic protein-induced protein 1; Reg1; Regnase-1; RGD:1306776}; RGD1306776; ribonuclease ZC3H12A; RP3-423B22.1; zc3h12a; zc3h12a {ECO:0000312; Zinc finger CCCH domain-containing protein 12A; zinc finger CCCH type containing 12A; zinc finger CCCH-type containing 12A | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 80149 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q5D1E8 | |
| ZC3H12A | |
| KLH conjugated synthetic peptide between 331-358 amino acids from the central region of human ZC12A. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction