missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ZC3H11A Antibody, Novus Biologicals™

Product Code. 18343319 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343319 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18343319 Supplier Novus Biologicals™ Supplier No. H00009877B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

ZC3H11A Polyclonal antibody specifically detects ZC3H11A in Human,Rat samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3H11A
Applications Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunocytochemistry/ Immunofluorescence
Formulation PBS (pH 7.4)
Gene Accession No. NP_055642.3
Gene Alias DKFZp686D03108, DKFZp686F14109, DKFZp781G2455, EC 4.1.1.31, EC 4.2.1.11, KIAA0663zinc finger CCCH-type domain containing 11A, ZC3HDC11A, zinc finger CCCH domain-containing protein 11A, zinc finger CCCH-type containing 11A
Host Species Mouse
Immunogen ZC3H11A (NP_055642.3, 1 a.a. - 810 a.a.) full-length human protein. MPNQGEDCYFFFYSTCTKGDSCPFRHCEAAIGNETVCTLWQEGRCFRQVCRFRHMEIDKKRSEIPCYWENQPTGCQKLNCAFHHNRGRYVDGLFLPPSKTVLPTVPESPEEEVKASQLSVQQNKLSVQSNPSPQLRSVMKVESSENVPSPTHPPVVINAADDDEDDDDQFSEEGDETKTPTLQPTPEVHNGLRVTSVRKPAVNIKQGECLNFGIKTLEEIKSKKMKEKSKKQGEGSSGVSSLLLHPEPVPGPEKENVRTVVRTVTLSTKQGEEPLVRLSLTERLGKRKFSAGGDSDPPLKRSLAQRLGKKVEAPETNIDKTPKKAQVSKSLKERLGMSADPDNEDATDKVNKVGEIHVKTLEEILLERASQKRGELQTKLKTEGPSKTDDSTSGARSSSTIRIKTFSEVLAEKKHRQQEAERQKSKKDTTCIKLKIDSEIKKTVVLPPIVASRGQSEEPAGKTKSMQEVHIKTLEEIKLEKALRVQQSSESSTSSPSQHEATPGARRLLRITKRTGMKEEKNLQEGNEVDSQSSIRTEAKEASGETTGVDITKIQVKRCETMREKHMQKQQEREKSVLTPLRGDVASCNTQVAEKPVLTAVPGITRHLTKRLPTKSSQKVEVETSGIGDSLLNVKCAAQTLEKRGKAKPKVNVKPSVVKVVSSPKLAPKRKAVEMHAAVIAAVKPLSSSSVLQEPPAKKAAVAVVPLVSEDKSVTVPEAENPRDSLVLPPTQSSSDSSPPEVSGPSSSQMSMKTRRLSSASTGKPPLSVEDDFEKLIWEISGGKLEAEIDLDPGKDEDDLLLELSEMIDS
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 9877
Target Species Human, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.