missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H11A Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74650AF405
This item is not returnable.
View return policy
Description
ZC3H11A Polyclonal antibody specifically detects ZC3H11A in Human samples. It is validated for Western Blot
Specifications
| ZC3H11A | |
| Polyclonal | |
| Western Blot | |
| DKFZp686D03108, DKFZp686F14109, DKFZp781G2455, EC 4.1.1.31, EC 4.2.1.11, KIAA0663zinc finger CCCH-type domain containing 11A, ZC3HDC11A, zinc finger CCCH domain-containing protein 11A, zinc finger CCCH-type containing 11A | |
| The immunogen recognized by this antibody maps to a region between residue 250 and 300 of human zinc finger CCCH-type containing 11A using the numbering given in entry NP_055642.3 (GeneID 9877). | |
| 0.1 mL | |
| DNA Repair | |
| 9877 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction