missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZC3H10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566478
This item is not returnable.
View return policy
Description
Immunogen sequence: RGGCCPPDGPH FESYEYSLAP PRGVECRLLE EENAMLRKRV EEL Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.
Specific regulator of miRNA biogenesis. Binds, via the C3H1-type zinc finger domains, to the binding motif 5'-GCAGCGC-3' on microRNA pri-MIR143 and negatively regulates the processing to mature microRNA.
Specifications
| ZC3H10 | |
| Polyclonal | |
| Unconjugated | |
| ZC3H10 | |
| AI790326; hypothetical protein zgc:77248; RPL41; SI:zC239J9.3; ZC3H10; ZC3HDC10; zgc:77248; Zinc finger CCCH domain-containing protein 10; zinc finger CCCH type containing 10; zinc finger CCCH-type containing 10; zinc finger CCCH-type domain containing 10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84872 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96K80 | |
| ZC3H10 | |
| Recombinant Human ZC3H10. Recombinant protein control fragment (Product #RP-107152). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction