missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20938
This item is not returnable.
View return policy
Description
ZC3H10 Polyclonal specifically detects ZC3H10 in Human samples. It is validated for Western Blot.
Specifications
| ZC3H10 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| FLJ14451, ZC3HDC10, zinc finger CCCH domain-containing protein 10, zinc finger CCCH-type containing 10, zinc finger CCCH-type domain containing 10 | |
| Rabbit | |
| 46 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Cow (97%), Mouse (95%), Rat (94%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.89mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q96K80 | |
| ZC3H10 | |
| Recombinant protein encompassing a sequence within the center region of human ZC3H10. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 84872 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction