missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZBTB9 Polyclonal Antibody

Description
A suggested positive control is mouse heart tissue lysate. PA5-21095 can be used with blocking peptide PEP-1209.
The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. ZBTB9, also known as ZNF919, is present in multiple tissues, and is highly expressed in the cytoplasm of the placenta. While little is known about ZBTB9, other ZBTB proteins, such as ZBTB4 bind methylated DNA and repress transcription. Another ZBTB proteins, ZBTB7A, has been implicated as a proto-oncogene whose overexpression contributes to malignancy in breast cancer, suggesting that ZBTB9 may act as a transcriptional repressor or be involved in tumorigenesis.
Specifications
Specifications
| Antigen | ZBTB9 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | ZBTB9 |
| Gene Accession No. | Q8CDC7, Q96C00 |
| Gene Alias | 3930402F13Rik; DASS-97D12.7; Zbtb9; zinc finger and BTB domain containing 9; zinc finger and BTB domain-containing protein 9; ZNF919 |
| Gene Symbols | ZBTB9 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?