Learn More
Invitrogen™ ZBTB9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140090
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: DAPRLTLPSV IEADAFEGLL QLIYSGRLRL PLDALPAHLL VASGLQMWQV VDQCSEILRE LETSGGGISA RGGNSYHALL STTSSTGGWC IRSSPFQT Highest antigen sequence indentity to the following orthologs: Mouse - 86%, Rat - 85%.
The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. ZBTB9, also known as ZNF919, is present in multiple tissues, and is highly expressed in the cytoplasm of the placenta. While little is known about ZBTB9, other ZBTB proteins, such as ZBTB4 bind methylated DNA and repress transcription. Another ZBTB proteins, ZBTB7A, has been implicated as a proto-oncogene whose overexpression contributes to malignancy in breast cancer, suggesting that ZBTB9 may act as a transcriptional repressor or be involved in tumorigenesis.
Specifications
| ZBTB9 | |
| Polyclonal | |
| Unconjugated | |
| ZBTB9 | |
| 3930402F13Rik; DASS-97D12.7; Zbtb9; zinc finger and BTB domain containing 9; zinc finger and BTB domain-containing protein 9; ZNF919 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 221504 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96C00 | |
| ZBTB9 | |
| Recombinant protein corresponding to Human ZBTB9. Recombinant protein control fragment (Product #RP-108954). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.