missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZBTB8B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571311
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Dog: 93%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Mouse: 79%; Rabbit: 86%; Rat: 86%.
ZBTB8B may be involved in transcriptional regulation.
Specifications
| ZBTB8B | |
| Polyclonal | |
| Unconjugated | |
| ZBTB8B | |
| AV220464; BC023839; BOZF1; putative zinc finger and BTB domain-containing protein 8B; RGD1562327; Zbtb8; ZBTB8B; Zbtb8l; zinc finger and BTB domain containing 8; zinc finger and BTB domain containing 8B; zinc finger and BTB domain-containing protein 8B; ZNF916B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 728116 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NAP8 | |
| ZBTB8B | |
| synthetic peptide directed towards the C-terminal region of human ZBTB8B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction