missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZBTB8A Polyclonal Antibody
GREENER_CHOICE

Product Code. 17249283 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17249283 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17249283 Supplier Invitrogen™ Supplier No. PA5116783

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Samples: U-87MG, PC-3, HeLa, Mouse lung, Mouse brain, Rat heart Immunogen sequence: HEPRKESIKK TKHLRLSQPS EVTHYKSSKR EVRTSDSSSH VSQSEEQAQI DAEMDSTPVG YQYGQGSDVT SKSFPDDLPR MRFKCPYCTH VVKRKADLKR HLRCHTGERP YPCQACGKRF SRLDHLSSHF RTIHQACKLI CRKCKRHVTD LTGQVVQEGT RRYRLCNECL AEFGIDSLPI DLEAEQHLMS PSDGDKDSRW H.

The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. ZBTB8, also known as BOZ-F1, is present in multiple tissues, and is highly expressed in the cytoplasm of the placenta. While little is known about ZBTB8, other ZBTB proteins, such as ZBTB4 bind methylated DNA and repress transcription. Another ZBTB proteins, ZBTB7A, has been implicated as a proto-oncogene whose overexpression contributes to malignancy in breast cancer, suggesting that ZBTB8 may act as a transcriptional repressor or be involved in tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB8A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.82 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ZBTB8A
Gene Accession No. B1WBU4, Q96BR9, Q9CWH1
Gene Alias 2410081M15Rik; AI480941; BOZF1; BOZ-F1; BTB/POZ and zinc-finger domain-containing factor; BTB/POZ and zinc-finger domains factor on chromosome 1; ZBTB8; Zbtb8a; zinc finger and BTB domain containing 8; zinc finger and BTB domain containing 8a; zinc finger and BTB domain-containing protein 8A; ZNF916A
Gene Symbols ZBTB8A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 200-400 of human ZBTB8A (NP_001035531).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 313049, 653121, 73680
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.