missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB8A Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76521JF549
This item is not returnable.
View return policy
Description
ZBTB8A Polyclonal antibody specifically detects ZBTB8A in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ZBTB8A | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 549 | |
| BOZ-F1, BOZF1BTB/POZ and zinc-finger domains factor on chromosome 1, BTB/POZ and zinc-finger domain-containing factor, FLJ90065, MGC17919, ZBTB8, zinc finger and BTB domain containing 8, zinc finger and BTB domain containing 8A, zinc finger and BTB domain-containing protein 8A, ZNF916A | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ZBTB8. The immunogen is located within the last 50 amino acids of ZBTB8. Amino Acid Squence: VDDSEEEEEKEIKPNIR | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 653121 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction