missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB7C Mouse anti-Human, Clone: PCRP-ZBTB7C-4E12, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | ZBTB7C |
|---|---|
| Clone | PCRP-ZBTB7C-4E12 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array |
| Applications | Flow Cytometry, Immunofluorescence, Peptide Array |
Description
ZBTB7C Monoclonal specifically detects ZBTB7C in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
| ZBTB7C | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunofluorescence, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| affected by papillomavirus DNA integration in ME180 cells protein 1, APM1, APM-1, B230208J24Rik, FLJ37907, ZBTB36, zinc finger and BTB domain containing 36, zinc finger and BTB domain containing 7C, zinc finger and BTB domain-containing protein 36, zinc finger and BTB domain-containing protein 7C, zinc finger protein 857C, ZNF857C | |
| Recombinant full-length human ZBTB7C protein | |
| Primary | |
| Store at 4°C. |
| PCRP-ZBTB7C-4E12 | |
| Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 201501 | |
| IgG2b | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title