missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB7B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11341-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a zinc finger-containing transcription factor that acts as a key regulator of lineage commitment of immature T-cell precursors. It is necessary and sufficient for commitment of CD4 lineage, while its absence causes CD8 commitment. It also functions as a transcriptional repressor of type I collagen genes. Alternatively spliced transcript variants have been found for this gene.Specifications
| ZBTB7B | |
| Polyclonal | |
| Unconjugated | |
| ZBTB7B | |
| c Krox, hcKrox, THPOK, ZBTB15, ZBTB7B, Zfp 67, ZFP67, Zinc finger protein 67 homolog, Zinc finger protein Th POK, ZNF857B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22724, 295248, 51043 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O15156, Q64321 | |
| ZBTB7B | |
| ZBTB7B Fusion Protein Ag1888 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction