missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ZBTB7B 293T Cell Transient Overexpression Lysate (Denatured) (T02)

Product Code. 16169552
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16169552 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16169552 Supplier Abnova™ Supplier No. H00051043T02.100uL
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human overexpression lysate

Specifications

Cell Line 293T
For Use With (Application) Western Blot
Description ZBTB7B 293T Cell Transient Overexpression Lysate(Denatured)
Gene Accession No. BC012070.1
Molecular Weight (g/mol) Theoretical M.W. 58kDa
Plasmid pXD-ZBTB7B full-length
Quality Control Testing Transient overexpression cell lysate was tested with anti-ZBTB7B antibody (Mfr. No. H00051043-B01P) by Western Blot.
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
compliance-icons
Product Identifier
  • 100UL ZBTB7B 293T Cell Transient Overexpression Ly
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: Mercaptoethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.