missing translation for 'onlineSavingsMsg'
Learn More

ZBTB7A/Pokemon Rabbit anti-Human, Clone: 3T2J1, Novus Biologicals™

Product Code. 18381367 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18381367 20 μg 20µL
18300843 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18381367 Supplier Novus Biologicals Supplier No. NBP31591420UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ZBTB7A/Pokemon Monoclonal antibody specifically detects ZBTB7A/Pokemon in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB7A/Pokemon
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3T2J1
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias DKFZp547O146, Factor binding IST protein 1, Factor that binds to inducer of short transcripts protein 1, FBI-1POK erythroid myeloid ontogenic factor, FBI1TTF-I-interacting peptide 21, HIV-1 inducer of short transcripts binding protein, Leukemia/lymphoma-related factor, LRFHIV-1 1st-binding protein 1, lymphoma related factor, MGC99631, pokemon, TIP21, ZBTB7zinc finger and BTB domain containing 7, zinc finger and BTB domain containing 7A, zinc finger and BTB domain containing 7A, HIV-1 inducer of short transcriptsbinding protein, zinc finger and BTB domain-containing protein 7A, Zinc finger protein 857A, ZNF857APOZ and Krueppel erythroid myeloid ontogenic factor
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 450-550 of human ZBTB7A/Pokemon (O95365).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 51341
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.