missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZBTB6 Polyclonal Antibody

Description
A suggested positive control is EL4 cell lysate. PA5-21086 can be used with blocking peptide PEP-1200.
The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. ZBTB6, also known as ZNF482, is highly expressed in brain. While little is known about ZBTB6, other ZBTB proteins, such as ZBTB4 bind methylated DNA and repress transcription. Another ZBTB family member, ZBTB5, also acts as a transcriptional repressor, and can interact with co-repressor histone deacetylase complexes such as BCoR, NCoR, and SMRT via its POZ domain, suggesting that ZBTB6 may also act as a transcriptional repressor.
Specifications
Specifications
| Antigen | ZBTB6 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | ZBTB6 |
| Gene Accession No. | Q15916, Q8K088 |
| Gene Alias | A830092L04Rik; ZBTB6; Zfp482; ZID; zinc finger and BTB domain containing 6; zinc finger and BTB domain-containing protein 6; Zinc finger protein 482; zinc finger protein with interaction domain; ZNF482 |
| Gene Symbols | ZBTB6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?