missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB5 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32099PE
This item is not returnable.
View return policy
Description
ZBTB5 Polyclonal antibody specifically detects ZBTB5 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| ZBTB5 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| KIAA0354, zinc finger and BTB domain containing 5, zinc finger and BTB domain-containing protein 5 | |
| The immunogen this antibody was made to, maps to a region between residue 200 to 250 of human Zinc Finger and BTB Domain Containing 5 using the numbering given in entry NP_055687.1 (GeneID 9925). | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 9925 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction