missing translation for 'onlineSavingsMsg'
Learn More

ZBTB48 Antibody, Novus Biologicals™

Product Code. 18364468 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364468 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364468 Supplier Novus Biologicals Supplier No. H00003104B01P50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

ZBTB48 Polyclonal antibody specifically detects ZBTB48 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB48
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10-1:500
Formulation PBS (pH 7.4)
Gene Accession No. NP_005332.1
Gene Alias GLI-Kruppel family member HKR3, HKR3zinc finger and BTB domain-containing protein 48, Krueppel-related zinc finger protein 3, Protein HKR3, zinc finger and BTB domain containing 48, Zinc finger protein 855, ZNF855pp9964
Host Species Mouse
Immunogen ZBTB48 (NP_005332.1, 1 a.a. - 688 a.a.) full-length human protein. MDGSFVQHSVRVLQELNKQREKGQYCDATLDVGGLVFKAHWSVLACCSHFFQSLYGDGSGGSVVLPAGFAEIFGLLLDFFYTGHLALTSGNRDQVLLAARELRVPEAVELCQSFKPKTSVGQAAGGQSGLGPPASQNVNSHVKEPAGLEEEEVSRTLGLVPRDQEPRGSHSPQRPQLHSPAQSEGPSSLCGKLKQALKPCPLEDKKPEDCKVPPRPLEAEGAQLQGGSNEWEVVVQVEDDGDGDYMSEPEAVLTRRKSNVIRKPCAAEPALSAGSLAAEPAENRKGTAVPVECPTCHKKFLSKYYLKVHNRKHTGEKPFECPKCGKCYFRKENLLEHEARNCMNRSEQVFTCSVCQETFRRRMELRVHMVSHTGEMPYKCSSCSQQFMQKKDLQSHMIKLHGAPKPHACPTCAKCFLSRTELQLHEAFKHRGEKLFVCEECGHRASSRNGLQMHIKAKHRNERPHVCEFCSHAFTQKANLNMHLRTHTGEKPFQCHLCGKTFRTQASLDKHNRTHTGERPFSCEFCEQRFTEKGPLLRHVASRHQEGRPHFCQICGKTFKAVEQLRVHVRRHKGVRKFECTECGYKFTRQAHLRRHMEIHDRVENYNPRQRKLRNLIIEDEKMVVVALQPPAELEVGSAEVIVESLAQGGLASQLPGQRLCAEESFTGPGVLEPSLIITAAVPEDCDT
Purification Method Immunogen affinity purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3104
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.