missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZBTB46 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA573021
This item is not returnable.
View return policy
Description
The amino acid sequence used as immunogen is 100% homologous in Chimpanzee and 88% homologous in Mouse.
ZBTB46 (Zinc Finger And BTB Domain Containing 46) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include nucleic acid binding. ZBTB46 also functions as a transcriptional repressor for PRDM1.
Specifications
| ZBTB46 | |
| Polyclonal | |
| Unconjugated | |
| ZBTB46 | |
| 2610019F01Rik; 4933406L05Rik; BTB (POZ) domain containing 4; BTB/POZ domain-containing protein 4; Btbd4; BTB-ZF protein expressed in effector lymphocytes; BZEL; dJ583P15.7; dJ583P15.8; RINZF; ZBTB46; zinc finger and BTB domain containing 46; zinc finger and BTB domain-containing protein 46; Zinc finger protein 340; ZNF340 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 140685 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 0.5 mg/mL | |
| PBS with 0.05% BSA and 0.05% sodium azide | |
| Q86UZ6 | |
| ZBTB46 | |
| Amino acids 62-87 of human RINZF protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction