missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB46 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£294.00 - £411.00
Specifications
| Antigen | ZBTB46 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18465141
|
Novus Biologicals
NBP1-88506-25ul |
25 μL |
£294.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18725823
|
Novus Biologicals
NBP1-88506 |
0.1 mL |
£411.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZBTB46 Polyclonal specifically detects ZBTB46 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ZBTB46 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| BTB (POZ) domain containing 4, BTB/POZ domain-containing protein 4, BTBD4, dJ583P15.7, dJ583P15.8, FLJ13502, RINZF, zinc finger and BTB domain containing 46, zinc finger and BTB domain-containing protein 46, Zinc finger protein 340, ZNF340 | |
| ZBTB46 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 140685 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:QACHDFIKAALDISIKSDASDELAEFEIGASSSSSTEALISAVMAGRSISPWLARRTSPANSSGDSAIASCHDGGSSYGKEDQEPKADGPDDVSSQPLWPGDVGYGPLRIKEEQVSPSQYGGSELPSAKDGAVQ | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title