missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB40 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20026-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody can recognize all isoforms of ZBTB40.
Accurate initiation of translation in eukaryotes is complex and requires many factors, some of which are composed of multiple subunits. The process is simpler in prokaryotes which have only three initiation factors (IF1, IF2, IF3). Two of these factors are conserved in eukaryotes: the homolog of IF1 is eIF1A and the homolog of IF2 is eIF5B. This gene encodes eIF5B. Factors eIF1A and eIF5B interact on the ribosome along with other initiation factors and GTP to position the initiation methionine tRNA on the start codon of the mRNA so that translation initiates accurately.Specifications
| ZBTB40 | |
| Polyclonal | |
| Unconjugated | |
| ZBTB40 | |
| KIAA0478, ZBTB40 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9923 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NUA8 | |
| ZBTB40 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction