missing translation for 'onlineSavingsMsg'
Learn More

ZBTB40 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18424578 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424578 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424578 Supplier Novus Biologicals Supplier No. NBP281868FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZBTB40 Polyclonal antibody specifically detects ZBTB40 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB40
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias KIAA0478ZNF923, MGC133098, zinc finger and BTB domain containing 40, zinc finger and BTB domain-containing protein 40
Host Species Rabbit
Immunogen ZBTB40 antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human ZBTB40. The immunogen is located within amino acids 180 - 230 of ZBTB40. Amino Acid Squence: PTAQESQRNAETPAETPTT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9923
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.