missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Description
ZBTB4 Polyclonal antibody specifically detects ZBTB4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | ZBTB4 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, ELISA 1:1000-1:5000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL, Immunohistochemistry-Paraffin 1:20-1:50 |
| Formulation | Whole antiserum |
| Gene Accession No. | Q9P1Z0 |
| Gene Alias | KAISO-L1, zinc finger and BTB domain containing 4 |
| Host Species | Rabbit |
| Immunogen | Polyclonal antibody raised in rabbit against human ZBTB4 (zinc finger and BTB domain containing 4), using four different KLH-conjugated synthetic peptides. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?