missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB39 Antibody (PCRP-ZBTB39-1A11), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08799AF594
This item is not returnable.
View return policy
Description
ZBTB39 Monoclonal antibody specifically detects ZBTB39 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array
Specifications
| ZBTB39 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| KIAA0352ZNF922, zinc finger and BTB domain containing 39, zinc finger and BTB domain-containing protein 39 | |
| Recombinant full-length human ZBTB39 protein (Uniprot: O15060) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence, Protein Array | |
| PCRP-ZBTB39-1A11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 9880 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction