missing translation for 'onlineSavingsMsg'
Learn More

ZBTB38 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18379624 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379624 100 μg 100µL
18388425 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18379624 Supplier Novus Biologicals Supplier No. NBP318641100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZBTB38 Polyclonal antibody specifically detects ZBTB38 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB38
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:100-1:1000
Formulation Whole antiserum
Gene Accession No. Q8NAP3
Gene Alias CIBZ, FLJ22332, FLJ31131, FLJ35036, zinc finger and BTB domain containing 38, zinc finger and BTB domain-containing protein 38, ZNF921
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against human ZBTB38 (zinc finger and BTB domain containing 38), using three KLH-conjugated synthetic peptides, two containing a sequence from the central part and one containing a sequence from the C-terminal part of the protein.
Purification Method Whole Antiserum
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253461
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Antisera
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.