missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZBTB38 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5149068
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
ZBTB38 is a methyl CpG binding protein that binds to DNA sequences containing a single methylated CpG, thereby acting as a transcriptional repressor. It has been found to be highly expressed in the brain and may be involved in the differentiation of late postmitotic neurons.
Specifications
| ZBTB38 | |
| Polyclonal | |
| Unconjugated | |
| ZBTB38 | |
| A930014K01Rik; CIBZ; CtBP-interacting BTB zinc finger protein; FLJ22332; FLJ31131; FLJ35036; PPP1R171; protein phosphatase 1, regulatory subunit 171; RGD1310136; si:ch73-160p19.2; UniProtKB:Q8BW71}; ZBTB38; zbtb38 {ECO:0000250; zenon; Zenon homolog; zinc finger and BTB domain containing 38; zinc finger and BTB domain-containing protein 38; Zinc finger protein expressed in neurons; zinc finger transcription factor Kaiso; ZNF921 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 253461 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8NAP3 | |
| ZBTB38 | |
| E. coli-derived recombinant human ZBTB38, Met1-Tyr232. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction