missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZBTB34 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571053
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 86%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.
ZBTB34 gene ontology annotations related to this gene include nucleic acid binding.
Specifications
| ZBTB34 | |
| Polyclonal | |
| Unconjugated | |
| ZBTB34 | |
| Gm349; KIAA1993; mKIAA1993; ZBTB34; zinc finger and BTB domain containing 34; zinc finger and BTB domain-containing protein 34; ZNF918 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 403341 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NCN2 | |
| ZBTB34 | |
| synthetic peptide directed towards the N-terminal region of human ZBTB34. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction