missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZBTB3 Polyclonal Antibody

Product Code. 12819232
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12819232 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12819232 Supplier Invitrogen™ Supplier No. PA521083

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is rat brain tissue lysate. PA5-21083 can be used with blocking peptide PEP-1197.

The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. While little is known about ZBTB3, the related protein ZBTB2 is thought to be phosphorylated in response to the DNA damage, probably by either ATM or ATR. Other ZBTB proteins, such as ZBTB4 and ZBTB38 bind methylated DNA and repress transcription, suggesting that ZBTB3 may also act as a transcription repressor.
TRUSTED_SUSTAINABILITY

Especificaciones

Antigen ZBTB3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ZBTB3
Gene Accession No. Q91X45, Q9H5J0
Gene Alias 4930563M09Rik; RGD1561140; ZBTB3; zinc finger and BTB domain containing 3; zinc finger and BTB domain-containing protein 3
Gene Symbols ZBTB3
Host Species Rabbit
Immunogen A 17 amino acid peptide near the carboxy terminus of human ZBTB3.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 499313, 75291, 79842
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.