missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB3 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
£436.00
Specifications
| Antigen | ZBTB3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 450 SE |
| Form | Purified |
Description
ZBTB3 Polyclonal antibody specifically detects ZBTB3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| ZBTB3 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ23392, zinc finger and BTB domain containing 3, zinc finger and BTB domain-containing protein 3 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ZBTB3. The immunogen is located within amino acids 510 - 560 of ZBTB3. Amino Acid Squence: RKAHNEDLAKRSKPDPE | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot | |
| mFluor Violet 450 SE | |
| Rabbit | |
| Signal Transduction | |
| 50mM Sodium Borate | |
| 79842 | |
| IgG | |
| Peptide affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title