missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB26 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20937
This item is not returnable.
View return policy
Description
ZBTB26 Polyclonal specifically detects ZBTB26 in Human samples. It is validated for Western Blot.
Specifications
| ZBTB26 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| KIAA1572, zinc finger and BTB domain containing 26, zinc finger and BTB domain-containing protein 26, Zinc finger protein 481bioref, zinc finger protein 483, Zinc finger protein Bioref, ZNF481 | |
| Rabbit | |
| 50 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1.03mg/mL | |
| Western Blot 1:1000-1:10000 | |
| Q9HCK0 | |
| ZBTB26 | |
| Recombinant protein encompassing a sequence within the center region of human ZBTB26. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 57684 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction