missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZBTB25 Monoclonal Antibody (2E8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524567
This item is not returnable.
View return policy
Description
Peptide Sequence: DTEPVELNCN FSFSRKRKMS CTICGHKFPR KSQLLEHMYT HKGKSYRYNR CQRFGNALAQ RFQPYCDSWS DVSLKSSRLS QEHLDLPCAL ESELTQENVD TILVE.
May be involved in transcriptional regulation. Classified as a putative transcription factor; human homolog is a DNA binding protein with two distantly spaced zinc fingers.
Specifications
| ZBTB25 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P24278 | |
| Zbtb25 | |
| ZBTB25 (NP_008908.2, 331 a.a. ~ 435 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunocytochemistry | |
| 2E8 | |
| Unconjugated | |
| Zbtb25 | |
| 2810462M08Rik; 2900064P18Rik; AI842128; C14orf51; KUP; Zbtb25; Zfp50; Zfp-50; zinc finger and BTB domain containing 25; zinc finger and BTB domain-containing protein 25; Zinc finger protein 46; zinc finger protein 50; zinc finger protein KUP; ZNF46 | |
| Mouse | |
| Protein A | |
| RUO | |
| 7597 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction