missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ZBTB25 293T Cell Transient Overexpression Lysate

Product Code. 18644011 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18644011 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18644011 Supplier Novus Biologicals™ Supplier No. H00007597T01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Quantity 0.1 mL
Content And Storage Store at -80C. Avoid freeze-thaw cycles.
Target ZBTB25
Synonym C14orf51, KUPchromosome 14 open reading frame 51, zinc finger and BTB domain containing 25, zinc finger and BTB domain-containing protein 25, Zinc finger protein 46, Zinc finger protein KUP, ZNF46zinc finger protein 46 (KUP)
Gene Symbol ZBTB25
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.