missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZBTB17 Recombinant Rabbit Monoclonal Antibody (HL1548)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547091
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Transcription factor that can function as an activator or repressor depending on its binding partners, and by targeting negative regulators of cell cycle progression. Plays a critical role in early lymphocyte development, where it is essential to prevent apoptosis in lymphoid precursors, allowing them to survive in response to IL7 and undergo proper lineage commitment. Has been shown to bind to the promoters of adenovirus major late protein and cyclin D1 and activate transcription. Required for early embryonic development during gastrulation.
Specifications
| ZBTB17 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q13105 | |
| ZBTB17 | |
| Recombinant protein encompassing a sequence within the center region of human Miz1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL1548 | |
| Unconjugated | |
| ZBTB17 | |
| AA589413; LP-1; Miz1; Miz-1; Myc-interacting zinc finger protein 1; Myc-interacting Zn finger protein-1; mZ13; pHZ-67; Polyomavirus late initiator promoter-binding protein; ZBTB17; Zfp100; Zfp-100; zinc finger and BTB domain containing 17; zinc finger and BTB domain-containing protein 17; Zinc finger protein 100; zinc finger protein 151; zinc finger protein 151 (pHZ-67); zinc finger protein 60; Zinc finger protein Z13; ZNF151; ZNF60 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7709 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction