missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZBTB14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562236
This item is not returnable.
View return policy
Description
Immunogen sequence: LRSDIFEEVL NYMYTAKISV KKEDVNLMMS SGQILGIRFL DKLCSQKRDV SSPDENNGQS KSKYCLKINR PIGDAADTQD DDVEEIGDQD DSP Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
Transcriptional activator of the dopamine transporter (DAT), binding it's promoter at the consensus sequence 5'-CCTGCACAGTTCACGGA-3'. Binds to 5'-d(GCC)(n)-3' trinucleotide repeats in promoter regions and acts as a repressor of the FMR1 gene. Transcriptional repressor of MYC and thymidine kinase promoters.
Specifications
| ZBTB14 | |
| Polyclonal | |
| Unconjugated | |
| ZBTB14 | |
| b2b1982Clo; Zbtb14; ZF5; ZFP161; zfp-161; ZFP161 zinc finger protein; Zfp5; Zfp-5; zinc finger and BTB domain containing 14; zinc finger and BTB domain-containing protein 14; Zinc finger protein 161; zinc finger protein 161 homolog; Zinc finger protein 478; zinc finger protein 5; Zinc finger protein 5 homolog; zinc finger protein homologous to Zfp161 in mouse; ZNF478 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7541 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43829 | |
| ZBTB14 | |
| Recombinant protein corresponding to Human ZBTB14. Recombinant protein control fragment (Product #RP-91702). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction