Learn More
ZBTB11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554992
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SGMGNDISAE DICAEDIPKH RQKVDQPLKD QENLVASTAK TDFGPDDDTY RSRLRQRSVN EGAYIRLHKG MEKKLQKRKA VPKSAVQQVA QKLVQRGKKM KQPKRDAKEN TEEASH Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 75%.
May be involved in transcriptional regulation.Specifications
ZBTB11 | |
Polyclonal | |
Unconjugated | |
ZBTB11 | |
9230110G02Rik; ZBTB11; zinc finger and BTB domain containing 11; zinc finger and BTB domain-containing protein 11; ZNF913; ZNF-U69274 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27107 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95625 | |
ZBTB11 | |
Recombinant protein corresponding to Human ZBTB11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only