Learn More
Invitrogen™ ZBTB10 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5113646
Product Code. 17161504
  • £436.00 / 100µL
Description
Immunogen sequence: SAWPPQPQPR QPPPPAPPAL QPPNGRGADE EVELEGLEPQ DLEASAGPAA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 93%.
This gene encodes a nuclear-localized protein that may be induced by p53 and regulates the cell cycle by inhibiting G1 to S phase progression. The encoded protein may interact with other cell cycle regulators.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ZBTB10
Polyclonal
Unconjugated
ZBTB10
4832414A18; AU021107; RINZF; RINZFC; ZBTB10; zinc finger and BTB domain containing 10; zinc finger and BTB domain-containing protein 10; Zinc finger protein RIN ZF; zinc finger protein RINZF
Rabbit
Affinity chromatography
RUO
65986
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q96DT7
ZBTB10
Synthetic peptide directed towards the N terminal region of human ZBTB10 (aa 36-85).