missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZBTB1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12818802
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12818802 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12818802 Supplier Invitrogen™ Supplier No. PA520653

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is HepG2 cell lysate. PA5-20653 can be used with blocking peptide PEP-0773.

ZBTB1 acts as a transcriptional repressor. Represses cAMP-responsive element (CRE)-mediated transcriptional activation. In addition, has a role in translesion DNA synthesis. Requires for UV-inducible RAD18 loading, PCNA monoubiquitination, POLH recruitment to replication factories and efficient translesion DNA synthesis. Plays a key role in the transcriptional regulation of T lymphocyte development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ZBTB1
Gene Accession No. Q91VL9, Q9Y2K1
Gene Alias 9430077A12; AI256369; brain specific protein 2; Bsg2; C430003J21Rik; KIAA0997; Zbtb1; zinc finger and BTB domain containing 1; zinc finger and BTB domain-containing protein 1; ZNF909
Gene Symbols ZBTB1
Host Species Rabbit
Immunogen A 14 amino acid peptide near the center of human ZBTB1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22890, 268564, 314246
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.